missing translation for 'onlineSavingsMsg'
Learn More

(4-Fluorobenzyl)triphenylphosphonium chloride, 98+%, Thermo Scientific™

Product Code. 11489822
Change view
Click to view available options
Quantity:
100 g
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11489822 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11489822 Supplier Thermo Scientific Alfa Aesar Supplier No. A10852.22
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3462-95-1
Molecular Formula C25H21ClFP
Molecular Weight (g/mol) 406.87
MDL Number MFCD00031637
InChI Key CBHDAHHYMRXLIP-UHFFFAOYSA-M
Synonym 4-fluorobenzyl triphenylphosphonium chloride, phosphonium, 4-fluorophenyl methyl triphenyl-, chloride, 4-fluorophenyl methyl triphenylphosphanium chloride, acmc-20a9k1, ksc495g9t, guljcsointwknf-uhfffaoysa-m, 4-fluorobenzyl triphenylphosphine chloride, p-fluorobenzyltriphenylphosphonium chloride, 4-fluorobenzyltriphenyl phosphonium chloride
PubChem CID 2733554
SMILES [Cl-].FC1=CC=C(C[P+](C2=CC=CC=C2)(C2=CC=CC=C2)C2=CC=CC=C2)C=C1

Specifications

Melting Point 310°C to 312°C
Quantity 100 g
Beilstein 4075331
IUPAC Name (4-fluorophenyl)methyl-triphenylphosphanium;chloride
Formula Weight 406.87
Percent Purity ≥98%
Chemical Name or Material (4-Fluorobenzyl)triphenylphosphonium chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.