accessibility menu, dialog, popup

UserName

CAS RN 3462-95-1

PFCl

CAS RN 3462-95-1

IUPAC Name: [(4-fluorophenyl)methyl]triphenylphosphanium chloride
Synonyms: [(4-fluorophenyl)methyl]triphenylphosphanium chloride
Molecular Weight (g/mol): 406.87
Molecular Formula: C25H21ClFP
InChi Key: CBHDAHHYMRXLIP-UHFFFAOYSA-M
SMILES: [Cl-].FC1=CC=C(C[P+](C2=CC=CC=C2)(C2=CC=CC=C2)C2=CC=CC=C2)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results