missing translation for 'onlineSavingsMsg'
Learn More

4-Fluoro-beta-nitrostyrene, 98%, Thermo Scientific™

Product Code. 15421899
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
This item is not returnable. View return policy

Product Code. 15421899

Brand: Thermo Scientific Alfa Aesar H64613.22

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
It is used as pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Store at 99-102°C. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 706-08-1
Molecular Formula C8H6FNO2
Molecular Weight (g/mol) 167.14
MDL Number MFCD00024824
InChI Key VRFSQVFSQAYHRU-AATRIKPKSA-N
Synonym 1-fluoro-4-2-nitrovinyl benzene, 4-fluoro-beta-nitrostyrene, e-1-fluoro-4-2-nitrovinyl benzene, trans-p-fluoro-beta-nitrostyrene, p-fluoro-beta-nitrostyrene, styrene, p-fluoro-beta-nitro, 1-4-fluorophenyl-2-nitroethylene, trans p-fluorophenyl-1 nitro-2 ethylene french, styrene, p-fluoro-beta-nitro-, e, 1-fluoro-4-e-2-nitrovinyl benzene
PubChem CID 5381680
SMILES [O-][N+](=O)\C=C\C1=CC=C(F)C=C1

Specifications

Melting Point 99°C to 102°C
Color Yellow
Quantity 100 g
UN Number UN2811
Beilstein 2208372
Solubility Information Slightly soluble in water.
IUPAC Name 1-fluoro-4-[(E)-2-nitroethenyl]benzene
Formula Weight 167.14
Percent Purity 98%
Physical Form Crystalline Solid
Chemical Name or Material 4-Fluoro-beta-nitrostyrene
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.