accessibility menu, dialog, popup

UserName

CAS RN 706-08-1

ONOF(E)

CAS RN 706-08-1

IUPAC Name: 1-fluoro-4-[(1E)-2-nitroethenyl]benzene
Synonyms: styrene, p-fluoro-β-nitro-
Molecular Weight (g/mol): 167.14
Molecular Formula: C8H6FNO2
InChi Key: VRFSQVFSQAYHRU-AATRIKPKSA-N
SMILES: [O-][N+](=O)\C=C\C1=CC=C(F)C=C1
Physical Form 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results