missing translation for 'onlineSavingsMsg'
Learn More

4-Fluoro-a-oxo-benzenepropanoic Acid, TRC

Product Code. 30402130
Change view
Click to view available options
Quantity:
25 mg
50 mg
250 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30402130 250 mg
30403166 25 mg
30389065 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30402130 Supplier Toronto Research Chemicals Supplier No. TRCF621550250MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7761-30-0
Molecular Formula C9H7O3F
Molecular Weight (g/mol) 182.15
Synonym (p-Fluorophenyl)pyruvic Acid, 4-Fluorophenylpyruvic Acid
SMILES OC(=O)C(=O)Cc1ccc(F)cc1

Specifications

Quantity 250 mg
InChI Formula InChI=1S/C9H7FO3/c10-7-3-1-6(2-4-7)5-8(11)9(12)13/h1-4H,5H2,(H,12,13)
Formula Weight 182.0379
Chemical Name or Material 4-Fluoro-a-oxo-benzenepropanoic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.