CAS RN 7761-30-0
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
4-Fluoro-a-oxo-benzenepropanoic Acid, TRC
CAS: 7761-30-0 Molecular Formula: C9H7O3F Molecular Weight (g/mol): 182.15 Synonym: (p-Fluorophenyl)pyruvic Acid,4-Fluorophenylpyruvic Acid SMILES: OC(=O)C(=O)Cc1ccc(F)cc1