missing translation for 'onlineSavingsMsg'
Learn More

4-Fluoro-3-hydroxybenzoic acid, 97%, Thermo Scientific™

Product Code. 11905011 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11905011 1 g 1g
11915011 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11905011 Supplier Thermo Scientific Acros Supplier No. 442030010
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 51446-31-2
Molecular Formula C7H4FO3
Molecular Weight (g/mol) 155.11
MDL Number MFCD00269641
InChI Key QATKOZUHTGAWMG-UHFFFAOYSA-M
Synonym benzoic acid, 4-fluoro-3-hydroxy, 4-fluoro-3-hydroxybenzoicacid, 4-fluoro-3-hydroxy-benzoic acid, 3-hydroxy-4-fluorobenzoic acid, pubchem3494, ksc497o4f, 3-hydroxy-4-fluoro-benzoic acid, 4-fluoro-3-hydroxy benzoic acid, timtec-bb sbb008505, buttpark 34\01-10
PubChem CID 593635
IUPAC Name 4-fluoro-3-hydroxybenzoic acid
SMILES OC1=CC(=CC=C1F)C([O-])=O

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 215.0°C to 217.0°C
Color Brown to White
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 156.11
Percent Purity 97%
Physical Form Crystalline Powder or Powder
Chemical Name or Material 4-Fluoro-3-hydroxybenzoic acid
Show More Show Less
compliance-icons
Product Identifier
  • 4-Fluoro-3-hydroxybenzoic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.