accessibility menu, dialog, popup

UserName

CAS RN 51446-31-2

OHFOO

CAS RN 51446-31-2

IUPAC Name: 4-fluoro-3-hydroxybenzoate
Synonyms: 4-fluoro-3-hydroxybenzoate
Molecular Weight (g/mol): 155.11
Molecular Formula: C7H4FO3
InChi Key: QATKOZUHTGAWMG-UHFFFAOYSA-M
SMILES: OC1=CC(=CC=C1F)C([O-])=O
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results