missing translation for 'onlineSavingsMsg'
Learn More

4'-Fluoro-3-biphenylmethylamine hydrochloride, 99%, Thermo Scientific™

Product Code. 11429450
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11429450 1 g 1g
11419450 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11429450 Supplier Thermo Scientific Alfa Aesar Supplier No. H50298.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1195901-44-0
Molecular Formula C13H13ClFN
Molecular Weight (g/mol) 237.702
MDL Number MFCD02089423
InChI Key ZYGOVJWOSPPYEB-UHFFFAOYSA-N
Synonym 3-4-fluorophenyl phenyl methylaminehcl, 4'-fluoro-3-biphenylmethylamine hydrochloride, 1-4'-fluorobiphenyl-3-yl methylamine hydrochloride, 3-4-fluorophenyl phenyl methanamine hydrochloride, 3-4-fluorophenyl phenyl methylamine, hcl, 1-4-fluorobiphenyl-3-yl methylamine hydrochloride, 4'-fluorobiphenyl-3-methylamine hydrochloride, 1-4'-fluoro 1,1'-biphenyl-3-yl methanamine-hydrogen chloride 1/1, 1-4'-fluoro-1,1'-biphenyl-3-yl methanamine hydrochloride
PubChem CID 17998850
IUPAC Name [3-(4-fluorophenyl)phenyl]methanamine;hydrochloride
SMILES C1=CC(=CC(=C1)CN)C2=CC=C(C=C2)F.Cl

Specifications

Quantity 1 g
Formula Weight 237.71
Percent Purity 99%
Chemical Name or Material 4'-Fluoro-3-biphenylmethylamine hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.