accessibility menu, dialog, popup

UserName

CAS RN 1195901-44-0

NH2FHCl

CAS RN 1195901-44-0

IUPAC Name: 1-{4'-fluoro-[1,1'-biphenyl]-3-yl}methanamine hydrochloride
Synonyms: 1-{4'-fluoro-[1,1'-biphenyl]-3-yl}methanamine hydrochloride
Molecular Weight (g/mol): 237.7
Molecular Formula: C13H13ClFN
InChi Key: ZYGOVJWOSPPYEB-UHFFFAOYSA-N
SMILES: Cl.NCC1=CC=CC(=C1)C1=CC=C(F)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results