missing translation for 'onlineSavingsMsg'
Learn More

4-Fluoro-2-methylbenzoic acid, 99%, Thermo Scientific™

Product Code. 10081504 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10081504 5 g 5g
10284864 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10081504 Supplier Thermo Scientific Acros Supplier No. 399300050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 321-21-1
Molecular Formula C8H7FO2
Molecular Weight (g/mol) 154.14
MDL Number MFCD04115880
InChI Key KDXOONIQRUZGSY-UHFFFAOYSA-N
Synonym 4-fluoro-o-toluic acid, 2-methyl-4-fluorobenzoic acid, 4-fluoro-2-methylbenzoicacid, 4-fluoro-2-methyl-benzoic acid, benzoic acid, 4-fluoro-2-methyl, pubchem1327, 2-carboxy-5-fluorotoluene, acmc-209hr6, 4-fluoro-2-methyl benzoicacid, rarechem al be 0506
PubChem CID 3860639
IUPAC Name 4-fluoro-2-methylbenzoic acid
SMILES CC1=C(C=CC(=C1)F)C(=O)O

Specifications

Melting Point 168.0°C to 172.0°C
Assay Percent Range 98.5% min. (HPLC)
Packaging Glass bottle
Linear Formula FC6H3(CH3)COOH
Quantity 5 g
Solubility Information Solubility in water: insoluble
Formula Weight 154.14
Percent Purity 99%
Chemical Name or Material 4-Fluoro-2-methylbenzoic acid
compliance-icons
Product Identifier
  • 4-Fluoro-2-methylbenzoic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.