accessibility menu, dialog, popup

UserName

CH3FHOO

CAS RN 321-21-1

IUPAC Name: 4-fluoro-2-methylbenzoic acid
Synonyms: 4-fluoro-2-methylbenzoic acid
Molecular Weight (g/mol): 154.14
Molecular Formula: C8H7FO2
InChi Key: KDXOONIQRUZGSY-UHFFFAOYSA-N
SMILES: CC1=CC(F)=CC=C1C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results