missing translation for 'onlineSavingsMsg'
Learn More

4-Dimethylamino-2-methoxybenzaldehyde, 98%, Thermo Scientific Chemicals

£68.90 - £269.00

Chemical Identifiers

CAS 84562-48-1
Molecular Formula C10H13NO2
Molecular Weight (g/mol) 179.219
MDL Number MFCD00151814
InChI Key HGDRXADJVGVGBC-UHFFFAOYSA-N
PubChem CID 291350
IUPAC Name 4-(dimethylamino)-2-methoxybenzaldehyde
SMILES CN(C)C1=CC(=C(C=C1)C=O)OC
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11465627
missing translation for 'viewDocuments' Promotion Details
Thermo Scientific Alfa Aesar
B23598.03
1 g
£68.90
1g
View alternative products
missing translation for 'toSaveUpto' 
»
Please to purchase this item. Need a web account? Register with us today!
11475627
missing translation for 'viewDocuments' Promotion Details
Thermo Scientific Alfa Aesar
B23598.06
5 g
£269.00
5g
View alternative products
missing translation for 'toSaveUpto' 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

4-Dimethylamino-2-methoxybenzaldehyde is used to produce 2,6-Dichloro-N'-(4-dimethylamino-2-methoxy-benzylidene)-benzene-1,4-diamine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Dimethylamino-2-methoxybenzaldehyde is used to produce 2,6-Dichloro-N′-(4-dimethylamino-2-methoxy-benzylidene)-benzene-1,4-diamine.

Solubility
Slightly soluble 0.32 g/L at 25°C.

Notes
Air sensitive. Store in cool, dry conditions in well sealed containers. Store away from oxidizing agents.
Specifications

Chemical Identifiers

84562-48-1
179.219
HGDRXADJVGVGBC-UHFFFAOYSA-N
4-(dimethylamino)-2-methoxybenzaldehyde
C10H13NO2
MFCD00151814
291350
CN(C)C1=CC(=C(C=C1)C=O)OC

Specifications

84562-48-1
C10H13NO2
2719181
4-dimethylamino-2-methoxybenzaldehyde, 2-methoxy-4-dimethylaminobenzaldehyde, 4-dimethylamino-2-methoxy-benzaldehyde, benzaldehyde, 4-dimethylamino-2-methoxy, 2-methoxy-4-dimethylamino benzaldehyde, benzaldehyde,4-dimethylamino-2-methoxy, 2-methoxy-4-n,n-dimethylamino benzaldehyde
HGDRXADJVGVGBC-UHFFFAOYSA-N
4-(dimethylamino)-2-methoxybenzaldehyde
291350
98%
58°C to 60°C
MFCD00151814
Air Sensitive
Slightly soluble 0.32g/L at 25°C.
CN(C)C1=CC(=C(C=C1)C=O)OC
179.219
179.22
4-Dimethylamino-2-methoxybenzaldehyde
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Certificates

RUO – Research Use Only