missing translation for 'onlineSavingsMsg'
Learn More

4-Dimethylamino-2-methoxybenzaldehyde, 98%

Product Code. 11465627
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11465627 1 g 1g
11475627 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11465627 Supplier Thermo Scientific Alfa Aesar Supplier No. B23598.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 84562-48-1 | C10H13NO2 | 179.219 g/mol

4-Dimethylamino-2-methoxybenzaldehyde is used to produce 2,6-Dichloro-N'-(4-dimethylamino-2-methoxy-benzylidene)-benzene-1,4-diamine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Dimethylamino-2-methoxybenzaldehyde is used to produce 2,6-Dichloro-N′-(4-dimethylamino-2-methoxy-benzylidene)-benzene-1,4-diamine.

Solubility
Slightly soluble 0.32 g/L at 25°C.

Notes
Air sensitive. Store in cool, dry conditions in well sealed containers. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 84562-48-1
Molecular Formula C10H13NO2
Molecular Weight (g/mol) 179.219
MDL Number MFCD00151814
InChI Key HGDRXADJVGVGBC-UHFFFAOYSA-N
PubChem CID 291350
IUPAC Name 4-(dimethylamino)-2-methoxybenzaldehyde
SMILES CN(C)C1=CC(=C(C=C1)C=O)OC

Specifications

Melting Point 58°C to 60°C
Quantity 1 g
Beilstein 2719181
Sensitivity Air Sensitive
Synonym 4-dimethylamino-2-methoxybenzaldehyde, 2-methoxy-4-dimethylaminobenzaldehyde, 4-dimethylamino-2-methoxy-benzaldehyde, benzaldehyde, 4-dimethylamino-2-methoxy, 2-methoxy-4-dimethylamino benzaldehyde, benzaldehyde,4-dimethylamino-2-methoxy, 2-methoxy-4-n,n-dimethylamino benzaldehyde
Solubility Information Slightly soluble 0.32g/L at 25°C.
Formula Weight 179.22
Percent Purity 98%
Chemical Name or Material 4-Dimethylamino-2-methoxybenzaldehyde

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.