missing translation for 'onlineSavingsMsg'
Learn More

4-(Diethylamino)benzoic acid, 99%, Thermo Scientific™

Product Code. 10106733 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
Packaging:
Glass bottle
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10106733 1 g Glass bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10106733 Supplier Thermo Scientific Acros Supplier No. 217730010

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5429-28-7
MDL Number MFCD00002538
InChI Key LNYTUARMNSFFBE-UHFFFAOYSA-N
Synonym 4-diethylamino benzoic acid, benzoic acid, 4-diethylamino, 4-n,n-diethylaminobenzoic acid, p-diethylamino benzoic acid, 4-diethylaminobenzoicacid, acmc-209lfa, 4-diethylamino benzoic, p-diethylaminobenzoic acid, 4-diethylamino benzoicacid
PubChem CID 79480
IUPAC Name 4-(diethylamino)benzoic acid
SMILES CCN(CC)C1=CC=C(C=C1)C(=O)O

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 192.0°C to 195.0°C
Color Green
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Molecular Formula C11H15NO2
Linear Formula (C2H5)2NC6H4CO2H
Quantity 1 g
Beilstein 14, 429
Molecular Weight (g/mol) 193.24
Formula Weight 193.24
Percent Purity 99%
Physical Form Fine Crystalline Powder
Chemical Name or Material 4-(Diethylamino)benzoic acid, 99%
Show More Show Less
compliance-icons
Product Identifier
  • 4-(Diethylamino)benzoic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.