accessibility menu, dialog, popup

UserName

CH3NCH3OHO

CAS RN 5429-28-7

IUPAC Name: 4-(diethylamino)benzoic acid
Synonyms: 4-(diethylamino)benzoic acid
Molecular Weight (g/mol): 193.25
Molecular Formula: C11H15NO2
InChi Key: LNYTUARMNSFFBE-UHFFFAOYSA-N
SMILES: CCN(CC)C1=CC=C(C=C1)C(O)=O
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results