missing translation for 'onlineSavingsMsg'
Learn More

4-(Di-n-propylsulfamoyl)benzoyl chloride, 96%, Thermo Scientific™

Product Code. 11450260
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11450260 250 mg 250mg
11460260 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11450260 Supplier Thermo Scientific Alfa Aesar Supplier No. H50703.MD
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 29171-72-0
Molecular Formula C13H18ClNO3S
Molecular Weight (g/mol) 303.80
MDL Number MFCD00121817
InChI Key LCQXGFTXKWEWQC-UHFFFAOYSA-N
Synonym 4-dipropylsulfamoyl benzoyl chloride, 4-dipropylamino sulfonyl benzene-1-carbonyl chloride, 4-n,n-dipropylsulfamoyl benzoyl chloride, benzoyl chloride,4-dipropylamino sulfonyl, acmc-1cejw, 4-dipropylsulfamoyl-benzoyl chloride, 4-di-n-propylsulfamoyl benzoyl chloride, 4-dipropylaminosulfonyl benzoyl chloride
PubChem CID 2735528
IUPAC Name 4-(dipropylsulfamoyl)benzoyl chloride
SMILES CCCN(CCC)S(=O)(=O)C1=CC=C(C=C1)C(Cl)=O

Specifications

Melting Point 59°C to 61°C
Quantity 250 mg
UN Number UN3261
Sensitivity Moisture sensitive
Formula Weight 303.81
Percent Purity 96%
Chemical Name or Material 4-(Di-n-propylsulfamoyl)benzoyl chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.