accessibility menu, dialog, popup

UserName

H3CNCH3SOOOCl

CAS RN 29171-72-0

IUPAC Name: 4-(dipropylsulfamoyl)benzoyl chloride
Synonyms: 4-(dipropylsulfamoyl)benzoyl chloride
Molecular Weight (g/mol): 303.8
Molecular Formula: C13H18ClNO3S
InChi Key: LCQXGFTXKWEWQC-UHFFFAOYSA-N
SMILES: CCCN(CCC)S(=O)(=O)C1=CC=C(C=C1)C(Cl)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results