missing translation for 'onlineSavingsMsg'
Learn More

4-Chlorophenyl isothiocyanate, 98%

Product Code. 11420603
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
This item is not returnable. View return policy

Product Code. 11420603

Brand: Thermo Scientific Alfa Aesar A12693.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 2131-55-7 | C7H4ClNS | 169.626 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2131-55-7
Molecular Formula C7H4ClNS
Molecular Weight (g/mol) 169.626
MDL Number MFCD00004810
InChI Key MZZVFXMTZTVUFO-UHFFFAOYSA-N
PubChem CID 16480
IUPAC Name 1-chloro-4-isothiocyanatobenzene
SMILES C1=CC(=CC=C1N=C=S)Cl

Specifications

Melting Point 43°C to 46°C
Boiling Point 135°C to 136°C (24 mmHg)
Quantity 5 g
Odor Pungent
UN Number UN2923
Beilstein 471610
Synonym 4-chlorophenyl isothiocyanate, p-chlorophenyl isothiocyanate, 1-chloro-4-isothiocyanato-benzene, benzene, 1-chloro-4-isothiocyanato, 4-chlorophenylisothiocyanate, isothiocyanic acid, p-chlorophenyl ester, 4-chlor-phenyl-isothiocyanat, isothiocyanic acid 4-chlorophenyl ester, isothiocyanic acid, 4-chlorophenyl ester, benzene,1-chloro-4-isothiocyanato
Formula Weight 169.64
Percent Purity 98%
Chemical Name or Material 4-Chlorophenyl isothiocyanate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.