accessibility menu, dialog, popup

UserName

CAS RN 2131-55-7

ClNCS

CAS RN 2131-55-7

IUPAC Name: 1-chloro-4-isothiocyanatobenzene
Synonyms: 1-chloro-4-isothiocyanatobenzene
Molecular Weight (g/mol): 169.63
Molecular Formula: C7H4ClNS
InChi Key: MZZVFXMTZTVUFO-UHFFFAOYSA-N
SMILES: ClC1=CC=C(C=C1)N=C=S
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (1)
  • (1)
  • (1)