missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Chloro-D-phenylalanine methyl ester hydrochloride, 98%, Thermo Scientific™
£15.77 - £207.00
Chemical Identifiers
CAS | 33965-47-8 |
---|---|
Molecular Formula | C10H13Cl2NO2 |
Molecular Weight (g/mol) | 250.12 |
MDL Number | MFCD00236920 |
InChI Key | GCBCWTWQAFLKJG-UHFFFAOYNA-N |
Synonym | 4-chloro-d-phenylalanine methyl ester hydrochloride, h-d-phe 4-cl ome.hcl, 4-chloro-d-phe-ome hcl, unii-k88egn008p, r-methyl 2-amino-3-4-chlorophenyl propanoate hydrochloride, d-methyl 4-chlorophenylalaninate hydrochloride, d-phenylalanine, 4-chloro-, methyl ester, hydrochloride, methyl 2r-2-amino-3-4-chlorophenyl propanoate hydrochloride, 4-chloro-d-phe-omecl, 4-chloro-d-phe-ome*hcl |
PubChem CID | 24720899 |
SMILES | Cl.COC(=O)C(N)CC1=CC=C(Cl)C=C1 |
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
33965-47-8 | |
250.12 | |
GCBCWTWQAFLKJG-UHFFFAOYNA-N | |
24720899 |
C10H13Cl2NO2 | |
MFCD00236920 | |
4-chloro-d-phenylalanine methyl ester hydrochloride, h-d-phe 4-cl ome.hcl, 4-chloro-d-phe-ome hcl, unii-k88egn008p, r-methyl 2-amino-3-4-chlorophenyl propanoate hydrochloride, d-methyl 4-chlorophenylalaninate hydrochloride, d-phenylalanine, 4-chloro-, methyl ester, hydrochloride, methyl 2r-2-amino-3-4-chlorophenyl propanoate hydrochloride, 4-chloro-d-phe-omecl, 4-chloro-d-phe-ome*hcl | |
Cl.COC(=O)C(N)CC1=CC=C(Cl)C=C1 |
Specifications
33965-47-8 | |
C10H13Cl2NO2 | |
4-chloro-d-phenylalanine methyl ester hydrochloride, h-d-phe 4-cl ome.hcl, 4-chloro-d-phe-ome hcl, unii-k88egn008p, r-methyl 2-amino-3-4-chlorophenyl propanoate hydrochloride, d-methyl 4-chlorophenylalaninate hydrochloride, d-phenylalanine, 4-chloro-, methyl ester, hydrochloride, methyl 2r-2-amino-3-4-chlorophenyl propanoate hydrochloride, 4-chloro-d-phe-omecl, 4-chloro-d-phe-ome*hcl | |
Cl.COC(=O)C(N)CC1=CC=C(Cl)C=C1 | |
250.12 | |
250.12 | |
Powder |
White | |
MFCD00236920 | |
GCBCWTWQAFLKJG-UHFFFAOYNA-N | |
methyl 2-amino-3-(4-chlorophenyl)propanoate hydrochloride | |
24720899 | |
98% | |
4-Chloro-D-phenylalanine methyl ester hydrochloride |
Safety and Handling
missing translation for 'tsca' : No
missing translation for 'storageNote' : 2° to 8°C
Recommended Storage : Keep cold