missing translation for 'onlineSavingsMsg'
Learn More

4-Chloro-D-phenylalanine methyl ester hydrochloride, 98%, Thermo Scientific™

Product Code. 15438566
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15438566 250 mg 250mg
15418566 1 g 1g
15428566 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15438566 Supplier Thermo Scientific Alfa Aesar Supplier No. H63873.MD

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 33965-47-8
Molecular Formula C10H13Cl2NO2
Molecular Weight (g/mol) 250.12
MDL Number MFCD00236920
InChI Key GCBCWTWQAFLKJG-UHFFFAOYNA-N
Synonym 4-chloro-d-phenylalanine methyl ester hydrochloride, h-d-phe 4-cl ome.hcl, 4-chloro-d-phe-ome hcl, unii-k88egn008p, r-methyl 2-amino-3-4-chlorophenyl propanoate hydrochloride, d-methyl 4-chlorophenylalaninate hydrochloride, d-phenylalanine, 4-chloro-, methyl ester, hydrochloride, methyl 2r-2-amino-3-4-chlorophenyl propanoate hydrochloride, 4-chloro-d-phe-omecl, 4-chloro-d-phe-ome*hcl
PubChem CID 24720899
SMILES Cl.COC(=O)C(N)CC1=CC=C(Cl)C=C1

Specifications

Color White
IUPAC Name methyl 2-amino-3-(4-chlorophenyl)propanoate hydrochloride
Quantity 250 mg
Formula Weight 250.12
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 4-Chloro-D-phenylalanine methyl ester hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.