missing translation for 'onlineSavingsMsg'
Learn More

4-Chloro-3-methylphenyl isocyanate, 98%, Thermo Scientific™

Product Code. 11310436
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
1g
This item is not returnable. View return policy

Product Code. 11310436

Brand: Thermo Scientific Alfa Aesar H31099.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
It is employed as a intermediates for pharmaceutical.

Solubility
Hydrolyzes with water.

Notes
Moisture Sensitive, store away from water/moisture. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store under dry inert gas.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 51488-20-1
Molecular Formula C8H6ClNO
Molecular Weight (g/mol) 167.59
MDL Number MFCD11101408
InChI Key UKTKKMZDESVUEE-UHFFFAOYSA-N
PubChem CID 11400996
SMILES CC1=CC=C(C=C1Cl)N=C=O

Specifications

Quantity 1 g
Refractive Index 1.558
UN Number UN2206
Synonym 4-chloro-3-methylphenyl isocyanate, 4-chloro-3-methylphenylisocyanate, 2-chloro-5-isocyanato toluene, benzene,1-chloro-4-isocyanato-2-methyl, 1-chloranyl-4-isocyanato-2-methyl-benzene, 3-methyl-4-chlorophenyl isocyanate, benzene, 1-chloro-4-isocyanato-2-methyl
Solubility Information Hydrolyzes with water.
IUPAC Name 2-chloro-4-isocyanato-1-methylbenzene
Formula Weight 167.6
Percent Purity 98%
Chemical Name or Material 4-Chloro-3-methylphenyl isocyanate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.