accessibility menu, dialog, popup

UserName

CAS RN 51488-20-1

CH3ClNCO

CAS RN 51488-20-1

IUPAC Name: 2-chloro-4-isocyanato-1-methylbenzene
Synonyms: 2-chloro-4-isocyanato-1-methylbenzene
Molecular Weight (g/mol): 167.59
Molecular Formula: C8H6ClNO
InChi Key: UKTKKMZDESVUEE-UHFFFAOYSA-N
SMILES: CC1=CC=C(C=C1Cl)N=C=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results