missing translation for 'onlineSavingsMsg'
Learn More

4-Chloro-3-methylbenzeneboronic acid, 98%

Product Code. 11490767
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11490767 1 g 1g
11400777 5 g 5g
11410777 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11490767 Supplier Thermo Scientific Alfa Aesar Supplier No. B23179.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 161950-10-3 | C7H8BClO2 | 170.40 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 161950-10-3
Molecular Formula C7H8BClO2
Molecular Weight (g/mol) 170.40
MDL Number MFCD02683104
InChI Key UZDPQDBLCJDUAX-UHFFFAOYSA-N
Synonym 4-chloro-3-methylbenzeneboronic acid, 4-chloro-3-methylphenyl boronic acid, 4-chloro-m-tolueneboronic acid, 4-chloro-m-tolylboronic acid, 4-chloro-3-methyl-phenyl boronic acid, boronic acid, 4-chloro-3-methylphenyl, boronic acid, b-4-chloro-3-methylphenyl, acmc-209xzw, 4-chloro-3-methylphenylboronicacid, 3-methyl-4-chlorophenylboronic acid
PubChem CID 3786632
IUPAC Name (4-chloro-3-methylphenyl)boronic acid
SMILES CC1=C(Cl)C=CC(=C1)B(O)O

Specifications

Melting Point 238°C to 242°C
Quantity 1 g
Formula Weight 170.4
Percent Purity 98%
Chemical Name or Material 4-Chloro-3-methylbenzeneboronic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.