CAS RN 161950-10-3
CAS RN 161950-10-3
IUPAC Name:
(4-chloro-3-methylphenyl)boronic acid
Synonyms:
4-chloro-3-methylphenylboronic acid
Molecular Weight (g/mol):
170.4
Molecular Formula:
C7H8BClO2
InChi Key:
UZDPQDBLCJDUAX-UHFFFAOYSA-N
SMILES:
CC1=C(Cl)C=CC(=C1)B(O)O
1
–
1
of
1
results
1
4-Chloro-3-methylbenzeneboronic acid, 98%
CAS: 161950-10-3 Molecular Formula: C7H8BClO2 Molecular Weight (g/mol): 170.40 MDL Number: MFCD02683104 InChI Key: UZDPQDBLCJDUAX-UHFFFAOYSA-N Synonym: 4-chloro-3-methylbenzeneboronic acid,4-chloro-3-methylphenyl boronic acid,4-chloro-m-tolueneboronic acid,4-chloro-m-tolylboronic acid,4-chloro-3-methyl-phenyl boronic acid,boronic acid, 4-chloro-3-methylphenyl,boronic acid, b-4-chloro-3-methylphenyl,acmc-209xzw,4-chloro-3-methylphenylboronicacid,3-methyl-4-chlorophenylboronic acid PubChem CID: 3786632 IUPAC Name: (4-chloro-3-methylphenyl)boronic acid SMILES: CC1=C(Cl)C=CC(=C1)B(O)O