missing translation for 'onlineSavingsMsg'
Learn More

4-Carboxy-2-chlorobenzeneboronic acid, 97%, Thermo Scientific™

Product Code. 15439237
Click to view available options
Quantity:
5 g
Unit Size:
5g
This item is not returnable. View return policy

Product Code. 15439237

Brand: Thermo Scientific Alfa Aesar H52489.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
Reactant in the synthesis and characterization1, 3, 4-oxadiazole.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 851335-09-6
Molecular Formula C7H6BClO4
Molecular Weight (g/mol) 200.38
MDL Number MFCD07363763
InChI Key FYDHSNLXZRGWFU-UHFFFAOYSA-N
Synonym 4-carboxy-2-chlorophenylboronic acid, 2-chloro-4-carboxyphenylboronic acid, 4-carboxy-2-chlorobenzeneboronic acid, 3-chloro-4-dihydroxyboranyl benzoic acid, benzoic acid, 4-borono-3-chloro, acmc-209q4s, ksc497q8l, 4-borono-3-chloro benzoic acid, benzoic acid,4-borono-3-chloro, 4-carboxy-2-chlorophenyl boronic acid
PubChem CID 17750227
SMILES OB(O)C1=C(Cl)C=C(C=C1)C(O)=O

Specifications

Melting Point 208°C to 210°C
Quantity 5 g
IUPAC Name 4-borono-3-chlorobenzoic acid
Formula Weight 200.39
Percent Purity 97%
Chemical Name or Material 4-Carboxy-2-chlorobenzeneboronic acid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.