accessibility menu, dialog, popup

UserName

CAS RN 851335-09-6

BOOHClHOOH

CAS RN 851335-09-6

IUPAC Name: 3-chloro-4-(dihydroxyboranyl)benzoic acid
Synonyms: 3-chloro-4-(dihydroxyboranyl)benzoic acid
Molecular Weight (g/mol): 200.38
Molecular Formula: C7H6BClO4
InChi Key: FYDHSNLXZRGWFU-UHFFFAOYSA-N
SMILES: OB(O)C1=C(Cl)C=C(C=C1)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results