missing translation for 'onlineSavingsMsg'
Learn More

4-Bromobenzeneboronic acid N-methyldiethanolamine ester, 98%, Thermo Scientific™

Product Code. 11334755
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11334755 1 g 1g
11344755 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11334755 Supplier Thermo Scientific Alfa Aesar Supplier No. L17775.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
4-Bromobenzeneboronic acid N-methyldiethanolamine ester enables selective reaction of the bromo substituent with organometallic (e.g. lithium) derivatives.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 133468-58-3
Molecular Formula C11H15BBrNO2
Molecular Weight (g/mol) 283.96
MDL Number MFCD02093957
InChI Key OSYBCIKXYVRFGE-UHFFFAOYSA-N
Synonym 2-4-bromophenyl-6-methyl-1,3,6,2-dioxazaborocane, 4-bromobenzeneboronic acid n-methyldiethanolamine ester, 4-bromophenylboronic acid n-methyl diethanolamine cyclic ester, 4h-1,3,6,2-dioxazaborocine, 2-4-bromophenyl tetrahydro-6-methyl, akos brn-1101, acmc-20ek5u, 4-bromophenylboronic acid n-methyl diethanolami, 2-4-bromo-phenyl-6-methyl-1,3,6,2 dioxazaborocane, 2-4-bromophenyl-6-methyltetrahydro-4h-1,3,6,2-dioxazaborocine
PubChem CID 9971093
IUPAC Name 2-(4-bromophenyl)-6-methyl-1,3,6,2-dioxazaborocane
SMILES B1(OCCN(CCO1)C)C2=CC=C(C=C2)Br

Specifications

Melting Point 146°C to 149°C
Quantity 1 g
Beilstein 3320262
Solubility Information Insoluble in water.
Formula Weight 283.96
Percent Purity 98%
Chemical Name or Material 4-Bromobenzeneboronic acid N-methyldiethanolamine ester
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.