accessibility menu, dialog, popup

UserName

CAS RN 133468-58-3

CH3NOBOBr

CAS RN 133468-58-3

IUPAC Name: 2-(4-bromophenyl)-6-methyl-1,3,6,2-dioxazaborocane
Synonyms: 2-(4-bromophenyl)-6-methyl-1,3,6,2-dioxazaborocane
Molecular Weight (g/mol): 283.96
Molecular Formula: C11H15BBrNO2
InChi Key: OSYBCIKXYVRFGE-UHFFFAOYSA-N
SMILES: CN1CCOB(OCC1)C1=CC=C(Br)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results