missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Bromobenzeneboronic acid N-methyldiethanolamine ester, 98%, Thermo Scientific™
Click to view available options
Quantity:
250 mg
1 g
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications
4-Bromobenzeneboronic acid N-methyldiethanolamine ester enables selective reaction of the bromo substituent with organometallic (e.g. lithium) derivatives.
Solubility
Insoluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agents.
4-Bromobenzeneboronic acid N-methyldiethanolamine ester enables selective reaction of the bromo substituent with organometallic (e.g. lithium) derivatives.
Solubility
Insoluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agents.
Specifications
Specifications
Melting Point | 146°C to 149°C |
Quantity | 250 mg |
Beilstein | 3320262 |
Solubility Information | Insoluble in water. |
Formula Weight | 283.96 |
Percent Purity | 98% |
Chemical Name or Material | 4-Bromobenzeneboronic acid N-methyldiethanolamine ester |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'