missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-DL-phenylalanine, 98+%, Thermo Scientific Chemicals

Product Code. 11307865
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11307865 1 g 1g
11327865 250 mg 250mg
11317865 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11307865 Supplier Thermo Scientific Alfa Aesar Supplier No. L02833.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14091-15-7
Molecular Formula C9H10BrNO2
Molecular Weight (g/mol) 244.09
MDL Number MFCD00002599
InChI Key PEMUHKUIQHFMTH-UHFFFAOYSA-N
Synonym 2-amino-3-4-bromophenyl propanoic acid, p-bromo-dl-phenylalanine, 4-bromo-dl-phenylalanine, dl-4-br-phe-oh, 4-bromophenylalanine, 4-bromo-phenylalanine, h-dl-phe 4-br-oh, 2-amino-3-4-bromophenyl propionic acid, 2-amino-3-4-bromo-phenyl-propionic acid, p-br-phenylalanine
PubChem CID 85681
IUPAC Name 2-amino-3-(4-bromophenyl)propanoic acid
SMILES NC(CC1=CC=C(Br)C=C1)C(O)=O

Specifications

Melting Point ∽262°C (decomposition)
Beilstein 2212158
Quantity 1 g
Odor Odorless
Formula Weight 244.09
Percent Purity ≥98%
Chemical Name or Material 4-Bromo-DL-phenylalanine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.