accessibility menu, dialog, popup

UserName

NH2BrOHO

CAS RN 14091-15-7

IUPAC Name: (2S)-2-azaniumyl-3-(4-bromophenyl)propanoate
Synonyms: (2S)-2-ammonio-3-(4-bromophenyl)propanoate
Molecular Weight (g/mol): 244.09
Molecular Formula: C9H10BrNO2
InChi Key: PEMUHKUIQHFMTH-QMMMGPOBSA-N
SMILES: [NH3+][C@@H](CC1=CC=C(Br)C=C1)C([O-])=O
Percent Purity 
  • (3)
  • (2)
Quantity 
  • (2)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (3)
  • (2)
  • (2)
  • (2)
  • (1)
  • (2)