missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-3-(trifluoromethyl)benzenesulfonyl chloride, 97%, Thermo Scientific™

Product Code. p-4813077 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11915781 1 g 1g
11925781 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11915781 Supplier Thermo Scientific Acros Supplier No. 442380010

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 351003-47-9
Molecular Formula C7H3BrClF3O2S
Molecular Weight (g/mol) 323.52
InChI Key QCFIMBHKZZLZAE-UHFFFAOYSA-N
Synonym 4-bromo-3-trifluoromethyl benzenesulfonyl chloride, 4-bromo-3-trifluoromethyl benzene-1-sulfonyl chloride, 4-bromo-3-trifluoromethyl benzene sulphonyl chloride, bromo-3-trifluoromethyl benzenesulfonylchloride, 4-bromo-3-trifluoromethyl benzenesulfonylchloride, 4-bromo-3-trifluoromethyl phenyl chlorosulfone, 4-bromo-3-trifluoromethyl benzenesulphonylchloride, 4-bromo-3-trifluoromethyl-benzenesulfonyl chloride, 4-bromo-3-trifluoromethyl benzenesulphonyl chloride, benzenesulfonyl chloride, 4-bromo-3-trifluoromethyl
PubChem CID 2778606
IUPAC Name 4-bromo-3-(trifluoromethyl)benzenesulfonyl chloride
SMILES C1=CC(=C(C=C1S(=O)(=O)Cl)C(F)(F)F)Br

Specifications

Melting Point 30.0°C to 33.0°C
Density 1.8330g/mL
Assay Percent Range 96% min. (GC)
Packaging Glass Bottle
Quantity 1 g
Specific Gravity 1.833
Formula Weight 323.52
Percent Purity 97%
Chemical Name or Material 4-Bromo-3-(trifluoromethyl)benzenesulfonyl chloride
compliance-icons
Product Identifier
  • 4-Bromo-3-(trifluoromethyl)benzenesulfonyl chloride
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.