accessibility menu, dialog, popup

UserName

SOOClFFFBr

CAS RN 351003-47-9

IUPAC Name: 4-bromo-3-(trifluoromethyl)benzene-1-sulfonyl chloride
Synonyms: 4-bromo-3-(trifluoromethyl)benzenesulfonyl chloride
Molecular Weight (g/mol): 323.51
Molecular Formula: C7H3BrClF3O2S
InChi Key: QCFIMBHKZZLZAE-UHFFFAOYSA-N
SMILES: FC(F)(F)C1=C(Br)C=CC(=C1)S(Cl)(=O)=O
Molecular Weight (g/mol) 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results