missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-2-fluorobenzonitrile, 99+%

Product Code. 10379250 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
Unit Size:
1g
25g
5g
This item is not returnable. View return policy

Product Code. 10379250

Brand: Thermo Scientific Acros 290490050

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 105942-08-3 | C7H3BrFN | 200.01 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 105942-08-3
Molecular Formula C7H3BrFN
Molecular Weight (g/mol) 200.01
MDL Number MFCD00143264
InChI Key HGXWRDPQFZKOLZ-UHFFFAOYSA-N
Synonym 2-fluoro-4-bromobenzonitrile, 4-bromo-2-fluoro-benzonitrile, benzonitrile, 4-bromo-2-fluoro, 4-bromo-2-fluorobenzenecarbonitrile, 4-cyano-3-fluorobromobenzene, pubchem3221, 2-floro-4-bromobenzonitrile, 4-bromo-2-fluorobenzontrile, 4-bromo-6-fluorobenzonitrile, ksc490q4t
PubChem CID 736029
IUPAC Name 4-bromo-2-fluorobenzonitrile
SMILES FC1=CC(Br)=CC=C1C#N

Specifications

CAS Min % 99.0
CAS Max % 100.0
Melting Point 69°C to 72°C
Color White to Brown
Infrared Spectrum Authentic
Assay Percent Range 99% min. (GC)
Packaging Glass bottle
Linear Formula BrC6H3(F)CN
Quantity 5 g
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in most common organic solvents
Formula Weight 200.01
Percent Purity 99+%
Physical Form Crystalline Powder and/or Chunks
Chemical Name or Material 4-Bromo-2-fluorobenzonitrile
Show More Show Less
compliance-icons
Product Identifier
  • 4-Bromo-2-fluorobenzonitrile
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.