missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-2-fluorobenzonitrile, 99%

Product Code. p-7020954
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
This item is not returnable. View return policy

Product Code. 11494404

Brand: Thermo Scientific Alfa Aesar A13086.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 105942-08-3 | C7H3BrFN | 200.01 g/mol

4-Bromo-2-fluorobenzonitrile is used as a OLED intermediate, Pharmaceutical, electronic and chemical intermediate. It is used in the synthesis of heterocycles and liquid crystals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Bromo-2-fluorobenzonitrile is used as a OLED intermediate, Pharmaceutical, electronic and chemical intermediate. It is used in the synthesis of heterocycles and liquid crystals.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 105942-08-3
Molecular Formula C7H3BrFN
Molecular Weight (g/mol) 200.01
MDL Number MFCD00143264
InChI Key HGXWRDPQFZKOLZ-UHFFFAOYSA-N
Synonym 2-fluoro-4-bromobenzonitrile, 4-bromo-2-fluoro-benzonitrile, benzonitrile, 4-bromo-2-fluoro, 4-bromo-2-fluorobenzenecarbonitrile, 4-cyano-3-fluorobromobenzene, pubchem3221, 2-floro-4-bromobenzonitrile, 4-bromo-2-fluorobenzontrile, 4-bromo-6-fluorobenzonitrile, ksc490q4t
PubChem CID 736029
IUPAC Name 4-bromo-2-fluorobenzonitrile
SMILES FC1=CC(Br)=CC=C1C#N

Specifications

Melting Point 69°C to 72°C
Boiling Point 238°C to 239°C
Quantity 1 g
UN Number UN3439
Beilstein 4231143
Solubility Information Insoluble in water.
Formula Weight 200.01
Percent Purity 99%
Chemical Name or Material 4-Bromo-2-fluorobenzonitrile

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.