missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-2,3,5,6-tetrafluorobenzoyl chloride, 98%, Thermo Scientific Chemicals

Product Code. 11378956
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11378956 5 g 5g
11368956 1 g 1g
11388956 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11378956 Supplier Thermo Scientific Alfa Aesar Supplier No. L20153.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 122033-54-9
Molecular Formula C7BrClF4O
Molecular Weight (g/mol) 291.424
MDL Number MFCD03001173
InChI Key XVYIQLOULDERAO-UHFFFAOYSA-N
Synonym 4-bromo-2,3,5,6-tetrafluorobenzoylchloride, benzoyl chloride,4-bromo-2,3,5,6-tetrafluoro, acmc-20amnz, 4-bromanyl-2,3,5,6-tetrakis fluoranyl benzoyl chloride
PubChem CID 2783245
IUPAC Name 4-bromo-2,3,5,6-tetrafluorobenzoyl chloride
SMILES C1(=C(C(=C(C(=C1F)F)Br)F)F)C(=O)Cl

Specifications

Boiling Point 95°C to 100°C
Refractive Index 1.517
Quantity 5 g
UN Number UN3265
Beilstein 7295811
Sensitivity Moisture sensitive
Formula Weight 291.43
Percent Purity 98%
Chemical Name or Material 4-Bromo-2,3,5,6-tetrafluorobenzoyl chloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.