CAS RN 122033-54-9
CAS RN 122033-54-9
IUPAC Name:
4-bromo-2,3,5,6-tetrafluorobenzoyl chloride
Synonyms:
4-bromo-2,3,5,6-tetrafluorobenzoyl chloride
Molecular Weight (g/mol):
291.42
Molecular Formula:
C7BrClF4O
InChi Key:
XVYIQLOULDERAO-UHFFFAOYSA-N
SMILES:
FC1=C(F)C(C(Cl)=O)=C(F)C(F)=C1Br
1
–
1
of
1
results
1
4-Bromo-2,3,5,6-tetrafluorobenzoyl chloride, 98%, Thermo Scientific Chemicals
CAS: 122033-54-9 Molecular Formula: C7BrClF4O Molecular Weight (g/mol): 291.424 MDL Number: MFCD03001173 InChI Key: XVYIQLOULDERAO-UHFFFAOYSA-N Synonym: 4-bromo-2,3,5,6-tetrafluorobenzoylchloride,benzoyl chloride,4-bromo-2,3,5,6-tetrafluoro,acmc-20amnz,4-bromanyl-2,3,5,6-tetrakis fluoranyl benzoyl chloride PubChem CID: 2783245 IUPAC Name: 4-bromo-2,3,5,6-tetrafluorobenzoyl chloride SMILES: C1(=C(C(=C(C(=C1F)F)Br)F)F)C(=O)Cl