missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-1,2-difluorobenzene, 98+%

Product Code. 11453924
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11453924 100 g 100g
11433924 5 g 5g
11443924 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11453924 Supplier Thermo Scientific Alfa Aesar Supplier No. A11934.22
 Restricted Item
Estimated Shipment: 02-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 348-61-8 | C6H3BrF2 | 192.99 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 348-61-8
Molecular Formula C6H3BrF2
Molecular Weight (g/mol) 192.99
MDL Number MFCD00000304
InChI Key YMQPKONILWWJQG-UHFFFAOYSA-N
Synonym 1-bromo-3,4-difluorobenzene, 3,4-difluorobromobenzene, benzene, 4-bromo-1,2-difluoro, 3,4-difluorobromebenzene, 4-bromo-1,2-difluoro-benzene, 3,4-difluoro-1-bromobenzene, paragos 390209, fr bf de, pubchem2303, 3,4-difluorophenylbromide
PubChem CID 67675
IUPAC Name 4-bromo-1,2-difluorobenzene
SMILES FC1=CC=C(Br)C=C1F

Specifications

Melting Point -4°C
Density 1.707
Boiling Point 150°C to 151°C
Flash Point 33°C (91°F)
Refractive Index 1.505
Quantity 100 g
UN Number UN1993
Beilstein 1934811
Formula Weight 192.99
Percent Purity ≥98%
Chemical Name or Material 4-Bromo-1,2-difluorobenzene
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.