accessibility menu, dialog, popup

UserName

CAS RN 348-61-8

IUPAC Name: 4-bromo-1,2-difluorobenzene
Synonyms: 4-bromo-1,2-difluorobenzene
Molecular Weight (g/mol): 192.99
Molecular Formula: C6H3BrF2
InChi Key: YMQPKONILWWJQG-UHFFFAOYSA-N
SMILES: FC1=CC=C(Br)C=C1F
Boiling Point 
  • (5)
Percent Purity 
  • (3)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results