missing translation for 'onlineSavingsMsg'
Learn More

(+/-)-4-Boc-piperazine-2-carboxylic acid hydrate, 95%, Thermo Scientific™

Product Code. p-7029894
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11367229 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11367229 Supplier Thermo Scientific Alfa Aesar Supplier No. H27413.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
As active pharmaceutical drugs and intermediates possess different biological activities.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 128019-59-0
Molecular Formula C10H18N2O4
Molecular Weight (g/mol) 230.26
MDL Number MFCD01076218
InChI Key YRYAXQJXMBETAT-UHFFFAOYNA-N
Synonym 4-tert-butoxycarbonyl piperazine-2-carboxylic acid, 4-boc-piperazine-2-carboxylic acid, n-4-boc-2-piperazinecarboxylic acid, 4-n-boc-piperazine-2-carboxylic acid, 1-boc-piperazine-3-carboxylic acid, 4-n-boc-piperazine-2-carboxylicacid, piperazine-1,3-dicarboxylic acid 1-tert-butyl ester, 4-boc piperazine-2-carboxylic acid, 4-tert-butoxylcarbonyl-piperazine-2-carboxylic acid, 4-boc-2-piperazinecarboxylic acid
PubChem CID 5175855
IUPAC Name 4-[(2-methylpropan-2-yl)oxycarbonyl]piperazine-2-carboxylic acid
SMILES CC(C)(C)OC(=O)N1CCNC(C1)C(O)=O

Specifications

Quantity 1 g
Solubility Information Slightly soluble in water.
Formula Weight 230.26 (Anhydrous)
Percent Purity 95%
Chemical Name or Material (+/-)-4-Boc-piperazine-2-carboxylic acid hydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.