accessibility menu, dialog, popup

UserName

CH3H3CCH3OONNHOHO

CAS RN 128019-59-0

IUPAC Name: 4-[(tert-butoxy)carbonyl]piperazine-2-carboxylic acid
Synonyms: 4-(tert-butoxycarbonyl)piperazine-2-carboxylic acid
Molecular Weight (g/mol): 230.26
Molecular Formula: C10H18N2O4
InChi Key: YRYAXQJXMBETAT-UHFFFAOYNA-N
SMILES: CC(C)(C)OC(=O)N1CCNC(C1)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results