missing translation for 'onlineSavingsMsg'
Learn More

4-(Boc-amino)-N-Fmoc-L-phenylalanine, 95%

Product Code. 15498517
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15498517 250 mg 250mg
15478517 1 g 1g
15488517 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15498517 Supplier Thermo Scientific Alfa Aesar Supplier No. H52031.MD
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 174132-31-1 | C29H30N2O6 | 502.567 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 174132-31-1
Molecular Formula C29H30N2O6
Molecular Weight (g/mol) 502.567
MDL Number MFCD00237007
InChI Key KVUAOWDVYMUKPE-VWLOTQADSA-N
Synonym s-2-9h-fluoren-9-yl methoxy carbonyl amino-3-4-tert-butoxycarbonyl amino phenyl propanoic acid, fmoc-l-4-aminophe boc, fmoc-phe 4-nhboc-oh, fmoc-l-phe 4-nhboc-oh, 2s-3-4-tert-butoxycarbonyl amino phenyl-2-9h-fluoren-9-ylmethoxy carbonyl amino propanoic acid, fmoc-p-amino-phe boc-oh, ambotzfaa1741, fmoc-4-boc-amino-l-phenylalanine, s-fmoc-4-boc-amino phenylalanine, n-9h-fluorene-9-ylmethoxycarbonyl-4-tert-butoxycarbonylamino-l-phenylalanine
PubChem CID 2756117
IUPAC Name (2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-3-[4-[(2-methylpropan-2-yl)oxycarbonylamino]phenyl]propanoic acid
SMILES CC(C)(C)OC(=O)NC1=CC=C(C=C1)CC(C(=O)O)NC(=O)OCC2C3=CC=CC=C3C4=CC=CC=C24

Specifications

Quantity 250 mg
Formula Weight 502.57
Percent Purity 95%
Chemical Name or Material 4-(Boc-amino)-N-Fmoc-L-phenylalanine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.