accessibility menu, dialog, popup

UserName

H3CCH3H3COOHNOOHNHOO

CAS RN 174132-31-1

IUPAC Name: 3-(4-{[(tert-butoxy)carbonyl]amino}phenyl)-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)propanoic acid
Synonyms: 3-{4-[(tert-butoxycarbonyl)amino]phenyl}-2-{[(9H-fluoren-9-ylmethoxy)carbonyl]amino}propanoic acid
Molecular Weight (g/mol): 502.57
Molecular Formula: C29H30N2O6
InChi Key: KVUAOWDVYMUKPE-UHFFFAOYNA-N
SMILES: CC(C)(C)OC(=O)NC1=CC=C(CC(NC(=O)OCC2C3=CC=CC=C3C3=CC=CC=C23)C(O)=O)C=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results