missing translation for 'onlineSavingsMsg'
Learn More

4-Biphenylzinc bromide, 0.5M in THF, packaged under Argon in resealable ChemSeal™ bottles, Thermo Scientific™

Product Code. 15445355
Change view
Click to view available options
Quantity:
50 mL
Unit Size:
50mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15445355 50 mL 50mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15445355 Supplier Thermo Scientific Alfa Aesar Supplier No. H58355.AD
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 312693-44-0
Density 0.994
Molecular Formula C12H9BrZn
MDL Number MFCD02093780
Quantity 50 mL
UN Number UN3399
Sensitivity Air sensitive; May form a precipitate
Synonym 4-biphenylzinc bromide, 4-biphenylzinc bromide solution, 1,1'-biphenyl-4-yl bromo zinc, 4-biphenylzinc bromide 0.5 m in tetrahydrofuran, 4-biphenylzinc bromide solution, 0.5 m in thf, 4-biphenylzinc bromide, 0.5m in thf, packaged under argon in resealable chemseal bottles
InChI Key PJCKUYSUPQKJDR-UHFFFAOYSA-M
SMILES [Zn+]Br.C1=CC=C(C=C1)C1=CC=[C-]C=C1
IUPAC Name [1,1'-biphenyl]-4-ide; bromozincylium
Molecular Weight (g/mol) 298.49
PubChem CID 24722605
Formula Weight 298.49
Chemical Name or Material 4-Biphenylzinc bromide, Packaged under Argon in resealable ChemSeal™ bottles
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.