missing translation for 'onlineSavingsMsg'
Learn More

4-Benzyloxybenzeneboronic acid, 97%, Thermo Scientific™

Product Code. 11405787
Click to view available options
Quantity:
0.25 g
1 g
5 g
Unit Size:
0.25g
1g
5g
This item is not returnable. View return policy

Product Code. 11405787

Brand: Thermo Scientific Alfa Aesar B24351.01

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
It is used as electronic intermediate. It is also utilized in HPLC and NMR spectroscopy.

Notes
Store at room temperature. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 146631-00-7
Molecular Formula C13H13BO3
Molecular Weight (g/mol) 228.054
MDL Number MFCD01075705
InChI Key DMJHEIDWSIAXCS-UHFFFAOYSA-N
Synonym 4-benzyloxyphenylboronic acid, 4-benzyloxybenzeneboronic acid, 4-benzyloxy phenyl boronic acid, 4-benzyloxy phenylboronic acid, 4-benzyloxyphenyl boronic acid, 4-benzyl-oxy phenyl boranediol, boronic acid, 4-phenylmethoxy phenyl, 4-phenylmethoxyphenyl boronic acid, pubchem9561
PubChem CID 2734314
IUPAC Name (4-phenylmethoxyphenyl)boronic acid
SMILES B(C1=CC=C(C=C1)OCC2=CC=CC=C2)(O)O

Specifications

Melting Point 193°C to 198°C
Quantity 0.25 g
Formula Weight 228.06
Percent Purity 97%
Chemical Name or Material 4-Benzyloxybenzeneboronic acid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.