accessibility menu, dialog, popup

UserName

CAS RN 146631-00-7

HOBOHO

CAS RN 146631-00-7

IUPAC Name: [4-(benzyloxy)phenyl]boronic acid
Synonyms: 4-(benzyloxy)phenylboronic acid
Molecular Weight (g/mol): 228.05
Molecular Formula: C13H13BO3
InChi Key: DMJHEIDWSIAXCS-UHFFFAOYSA-N
SMILES: OB(O)C1=CC=C(OCC2=CC=CC=C2)C=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results