missing translation for 'onlineSavingsMsg'
Learn More

4-Benzyloxy-3-methoxyphenylacetonitrile, 97%, Thermo Scientific™

Product Code. 11354256
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11354256 250 mg 250mg
11344256 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11354256 Supplier Thermo Scientific Alfa Aesar Supplier No. L19331.MD
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1700-29-4
Molecular Formula C16H15NO2
Molecular Weight (g/mol) 253.301
MDL Number MFCD00016394
InChI Key KSOYPRFHKIOHMY-UHFFFAOYSA-N
Synonym 4-benzyloxy-3-methoxyphenylacetonitrile, 2-4-benzyloxy-3-methoxyphenyl acetonitrile, 4-benzyloxy-3-methoxybenzyl cyanide, 4-benzyloxy-3-methoxyphenyl acetonitrile, 2-3-methoxy-4-phenylmethoxyphenyl acetonitrile, benzeneacetonitrile, 3-methoxy-4-phenylmethoxy, acmc-209vuk, 4-benzyl-oxy-3-methoxyphenylacetonitrile, 3-methoxy-4-benzyloxyphenyl acetonitrile
PubChem CID 297963
IUPAC Name 2-(3-methoxy-4-phenylmethoxyphenyl)acetonitrile
SMILES COC1=C(C=CC(=C1)CC#N)OCC2=CC=CC=C2

Specifications

Melting Point 68°C to 70°C
Boiling Point 195°C (1 mmHg)
Quantity 250 mg
UN Number UN3439
Beilstein 2702492
Formula Weight 253.3
Percent Purity 97%
Chemical Name or Material 4-Benzyloxy-3-methoxyphenylacetonitrile
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.