accessibility menu, dialog, popup

UserName

H3CONO

CAS RN 1700-29-4

IUPAC Name: 2-[4-(benzyloxy)-3-methoxyphenyl]acetonitrile
Synonyms: 2-[4-(benzyloxy)-3-methoxyphenyl]acetonitrile
Molecular Weight (g/mol): 253.3
Molecular Formula: C16H15NO2
InChi Key: KSOYPRFHKIOHMY-UHFFFAOYSA-N
SMILES: COC1=CC(CC#N)=CC=C1OCC1=CC=CC=C1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results